Learn More
Description
Thermo Scientific™ Remel Autoclave Bag includes a Biohazard warning.
Specifications
Specifications
| Description | 200/Pk. |
| Autoclavable | Autoclavable |
| Quantity | 200 Pk. |
| Dimensions (L x W) | 890 x 635 mm |
| Length (Metric) | 89 cm |
| Width (Metric) | 63.5 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.