Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ BactiCard™ Strep
GSA_VA

Catalog No. r21112
Change view
Click to view available options
Quantity:
25 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21112 25 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R21112 Supplier Thermo Scientific™ Supplier No. R21112
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Thermo Scientific™ Remel BactiCard Strep is used for presumptive identification of streptococci and related genera using enzyme technology.

Thermo Scientific™ Remel BactiCard Strep is used for presumptive identification of streptococci and related genera using enzyme technology.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Control Sets Positive, Negative
Description BactiCard™ Strep
CE Marker Yes
Detectable Analytes Clumping Factor and Protein A
Type Bacticard for Streptococcus Testing
For Use With (Application) Enzyme Immunoassay Detection
Packaging Type 1 Kit
Quantity 25 Tests/Kit
Target Organism Class Staphylococcus aureus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.