Description
Remel BactiDrop India Ink is for microscopic examination of encapsulated microorganisms.
Ordering Alerts: Supplied in crushable ampules enclosed in a protective plastic sheath with a convenient dropper dispenser tip.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 50 Pk. |
| Volume (Metric) | 0.75 mL/Ampule |
| Product Type | India Ink |