Learn More
Description
Determine the ability of gram-negative bacilli to produce acetylmethylcarbinol (acetoin) from glucose fermentation using Thermo Scientific™ Remel™ Bactidrop™ Voges-Proskauer A Test. In 1898, Voges and Proskauer reported production of acetylmethylcarbinol from glucose fermentation by certain bacteria when grown in an appropriate medium such as MR-VP Broth, which is a modification of Clark and Lubs medium1,2. Later Barret made the VP test more sensitive by using 5% Alpha Naphthol (VP-A) to the medium for the detection of acetylmethylcarbinol3.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | 20°C to 25°C |
| Description | Bactidrop Vp-a Alpha Na |
| CE Marker | Yes |
| Detectable Analytes | Acetylmethylcarbinol |
| Quantity | 50 x 0.75 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
