Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ BactiDrop™ Voges-Proskauer B
GSA_VA

Catalog No. R21562 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 x 0.75 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21562 50 x 0.75 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R21562 Supplier Thermo Scientific™ Supplier No. R21562
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Determine the ability of gram-negative bacilli to produce acetylmethylcarbinol (acetoin) from glucose fermentation using Voges-Proskauer B Test.

Determine the ability of gram-negative bacilli to produce acetylmethylcarbinol (acetoin) from glucose fermentation using Thermo Scientific™ Remel™ Bactidrop™ Voges-Proskauer B Test. In 1898, Voges and Proskauer reported production of acetylmethylcarbinol from glucose fermentation by certain bacteria when grown in an appropriate medium such as MR-VP Broth which is a modification of Clark and Lubs medium 1,2. Later Barret made the VP test more sensitive by using 40% Potassium Hydroxide (VP-B) to the medium for the detection of acetylmethylcarbinol production3.

The BactiDrop™ Voges-Proskauer test is based on the detection of acetylmethylcarbinol production which is a precursor to 2,3-butanediol from glucose metabolism.Ready to use: Convenience of a prepared reagent.Ease of differentiation: VP positive organisms produce a pink color development while VP negative organisms produce no color.Rapid detection: Within 15 minutes.Glucose is metabolized to pyruvic acid from which the neutral products acetoin and 2.3-butanediol are formed. One molecule of acetoin is formed by the decarboxylation of two molecules of pyruvic acid. In the presence of oxygen and alkali, acetoin and 2,3-butanediol are oxidized to diacetyl which is the reactant for the pink color produced in Voges-Proskauer test. 40% potassium hydroxide aids in the absorption of CO2 to hasten the oxidation of acetoin to diacetyl.Used in conjunction with VP-A, Part No. R21560. For use with MRVP Broth.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 20°C to 25°C
Description BactiDrop Voges-Proskauer B
CE Marker Yes
Detectable Analytes Acetylmethylcarbinol
Quantity 50 x 0.75 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.