Learn More
Thermo Scientific™ BactiStaph™ Latex

Description
Rapidly identify Staphylococcus aureus and distinguish from other staphylococci by detecting clumping factor and protein A using the Thermo Scientific™ BactiStaph™ Latex.
Use with suspected S. aureus colonies. Prior to testing, suspect colonies should be Gram stained and a catalase test performed to verify that they are Staphylococcus.
Although Staphylococci are commonly found on the skin and in mucous membranes, they have been associated with many human and animal infections. S.aureus and other coagulase-positive Staphylococci have been identified as a cause of suppurative infections, food poisoning and toxic shock syndrome and have been isolated from nearly all anatomical sites.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Control Sets | Positive,Negative |
| Description | BactiStaph Latex |
| CE Marker | Yes |
| Detectable Analytes | S. aureus Coagulase, Protein A |
| Type | Staphylococcus aureus Test Kit |
| Includes | Latex reagent, positive control, negative control, disposable slides, stirring sticks |
| Material | Latex |
| Quantity | 150 Tests/Kit |
| Target Organism Class | Staphylococcus aureus |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.