Description
Thermo Scientific™ Remel BactiSwab Dry is supplied as a single plastic-shaft swab (contains no media).
Thermo Scientific™ Remel™ BactiSwab™ Dry is comprised of a sterile, rayon-tipped plastic shaft swab for use in collection and transport of a variety of sample to the laboratory for microbiological examination when dry swab collection is appropriate. The rayon swab is sterile and non-reactive and can be removed from the plastic sleeve without handling or contaminating the swab tip.
Ordering Alerts:
Each BactiSwab is supplied as one tube per envelope.
Each BactiSwab is supplied as a single plastic-shaft swab in an envelope. This product contains no media.
Specifications
Specifications
| Swab Configuration | Single Applicator |
| Description | Bacti-Swab Dry; Single plastic-shaft swab; 100/Pk. |
| For Use With (Application) | Healthcare |
| Shaft Material | Plastic |
| Tip Material | Rayon |