Learn More
Thermo Scientific™ Remel™ BactiSwab™ Gel Collection and Transport Swabs: Single Swab Amies Charcoal
Catalog No. R723040 Shop All Thermo Scientific Products
Change view
Click to view available options
Packaging:
50/Pk.
500/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
R723040 500/Pk.
R723045 50/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R723040 Supplier Thermo Scientific™ Supplier No. R723040
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel BactiSwab Gel Collection and Transport Swabs provide excellent recovery for a wide range of bacteria, including anaerobes.
Thermo Scientific™ Remel BactiSwab Gel Collection and Transport Swabs provide excellent recovery for a wide range of bacteria, including anaerobes.
Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing gel transport medium with or without charcoal.
  • Recommended for recovery of Neisseria gonorrhoeae
  • Single plastic shaft
  • Black cap
  • Round-bottom tube for easy racking and seamless molding to ensure strength and integrity
  • Exclusive zip-seal packaging to promote optimum media performance
  • Color-coded cap with protective overskirt
  • Inert, non-toxic rayon swab tip for good sample retrieval and absorption
TRUSTED_SUSTAINABILITY
Specifications
Format Tube
Media Type Amies Charcoal
Closure Color Black
Description Amies Charcoal
Packaging 500/Pk.
Quantity 1 Tube/Envelope
Shaft Material Plastic