Learn More
Thermo Scientific™ Remel™ BactiSwab™ Gel Collection and Transport Swabs: Single Swab Amies Charcoal
Shop All Thermo Scientific Products
Description
Thermo Scientific™ Remel BactiSwab Gel Collection and Transport Swabs provide excellent recovery for a wide range of bacteria, including anaerobes.
- Recommended for recovery of Neisseria gonorrhoeae
- Single plastic shaft
- Black cap
- Round-bottom tube for easy racking and seamless molding to ensure strength and integrity
- Exclusive zip-seal packaging to promote optimum media performance
- Color-coded cap with protective overskirt
- Inert, non-toxic rayon swab tip for good sample retrieval and absorption
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Format | Tube |
| Media Type | Amies Charcoal |
| Closure Color | Black |
| Description | Amies Charcoal |
| Packaging | 50/Pk. |
| Quantity | 1 Tube/Envelope |
| Shaft Material | Plastic |