Learn More
Description
Thermo Scientific™ Remel BactiSwab Gel: Wire Shaft Amies Clear is supplied with a mini-tip swab suitable for ear, nose, throat, eye, male urethral and pediatric specimen collection.
- Straight aluminum shaft
- Orange cap
- Round-bottom tube for easy racking and seamless molding to ensure strength and integrity
- Exclusive zip-seal packaging to promote optimum media performance
- Color-coded cap with protective overskirt
- Inert, non-toxic rayon swab tip for good sample retrieval and absorption
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Format | Tube |
| Media Type | Amies Clear |
| Closure Color | Orange |
| Description | Amies Clear |
| For Use With (Application) | Healthcare |
| Packaging | 50/Pk. |
| Quantity | 1 Tube/Envelope |
| Shaft Material | Wire |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
