Learn More
Thermo Scientific™ Blood Heart Infusion (BHI) Agar with Sheep Blood, Chloramphenicol and Cycloheximide
Shop All Thermo Scientific ProductsDescription
Cultivate a wide variety of microorganisms including bacteria, yeasts, and molds with the nutritious base medium Thermo Scientific™ Remel™ Blood Heart Infusion (BHI) Agar with Sheep Blood, Chloramphenicol and Cycloheximide1. Isolate fastidious fungi and grow Actinomyces2 using BHI Agar with 5-10% sheep blood. Pathogenic fungi and Nocardia asteroides can be isolated using optimal concentrations of chloramphenicol3. Cycloheximide inhibits certain Aspergillus spp. and zygomycetes.
- Chloramphenicol inhibits some gram-positive and gram-negative bacteria
- Cycloheximide inhibits rapid growing saprophytic fungi
Specifications
Specifications
| Description | BHI Agar w/Sheep Blood, Chloramphenicol, Cycloheximide |
| Format | Monoplate Deep Pour |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |