Learn More
Thermo Scientific™ Remel™ BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin, Cycloheximide
Shop All Thermo Scientific Products
Description
Thermo Scientific™ Remel BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin, Cycloheximide is used for selective isolation of fastidious fungi and yeast.
- Chloramphenicol inhibits some gram-positive and gram-negative microorganisms
- Gentamicin inhibits some gram-negative bacteria
- Cycloheximide inhibits rapid growing saprophytic fungi
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin, Cycloheximide |
| Format | Monoplate Deep Pour |
| Product Type | Agar |
| Packaging Type | 10/Pk. |
| Quantity | 10/Pk. |