Learn More
Thermo Scientific™ Remel™ BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin, Cycloheximide
Catalog No. R08078 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
Format:
F - Slants
Monoplate Deep Pour
Packaging Type:
10/Pk.
100/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Packaging Type
R08078 100/Pk. F - Slants 100/Pk.
R01148 10/Pk. Monoplate Deep Pour 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R08078 Supplier Thermo Scientific™ Supplier No. R08078
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin, Cycloheximide is used for selective isolation of fastidious fungi and yeast.
Thermo Scientific™ Remel BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin, Cycloheximide is used for selective isolation of fastidious fungi and yeast.
  • Chloramphenicol inhibits some gram-positive and gram-negative microorganisms
  • Gentamicin inhibits some gram-negative bacteria
  • Cycloheximide inhibits rapid growing saprophytic fungi
TRUSTED_SUSTAINABILITY
Specifications
Description BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin, Cycloheximide
Format F - Slants
Product Type Agar
Packaging Type 100/Pk.
Quantity 100/Pk.