Learn More
Thermo Scientific™ Remel™ BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin
GSA_VA
Catalog No. R01144 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
20/Pk.
Format:
F - slant
Monoplate Deep Pour
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R01144 10/Pk. Monoplate Deep Pour
R08072 20/Pk. F - slant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R01144 Supplier Thermo Scientific™ Supplier No. R01144
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
The Thermo Scientific™ Remel BHI Agar w/Sheep Blood, Chloramphenicol and Gentamicin is used for selective isolation of fastidious fungi and yeast.
The Thermo Scientific™ Remel BHI Agar w/Sheep Blood, Chloramphenicol and Gentamicin is used for selective isolation of fastidious fungi and yeast.
  • Chloramphenicol inhibits some gram-positive and gram-negative microorganisms
  • Gentamicin inhibits some gram-negative bacteria
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description BHI Agar w/Sheep Blood, Chloramphenicol, Gentamicin
Format Monoplate Deep Pour
Product Type Agar
Quantity 10/Pk.