Learn More
Thermo Scientific™ BHI Agar with Sheep Blood
GSA_VA
Catalog No. R01136
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
20/Pk.
Format:
20mm x 113mm Tube
Monoplate Deep Pour
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R01136 10/Pk. Monoplate Deep Pour
R01139 100/Pk. Monoplate Deep Pour
R08066 20/Pk. 20mm x 113mm Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R01136 Supplier Thermo Scientific™ Supplier No. R01136
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Cultivate a wide variety of microorganisms including bacteria, yeasts, and molds with this nutritious medium.
Cultivate a wide variety of microorganisms including bacteria, yeasts, and molds with nutritious Thermo Scientific™ Remel™ BHI Agar with Sheep Blood. Due to the addition of Sheep blood, the BHI Agar can be used for the isolation of dimorphic fungi which may grow poorly on non-enriched media.
Use BHI Agar with Sheep Blood for the cultivation of a wide variety of micro-organisms including pathogenic fungi.
  • Highly Nutritious: medium recommended for the enrichment of fastidious organisms.
  • Ready-to-use: convenience of prepared media.
BHI Agar w/Sheep Blood contains brain heart infusion, peptone, and dextrose which provide nitrogen, carbon, sulfur, vitamins and a carbohydrate source. Sheep blood provides essential growth factors for fastidious fungi. Yeast extracts are incorporated in the medium to serve as a growth stimulant.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description BHI Agar w/Sheep Blood
Format Monoplate Deep Pour
Product Type Agar Plate
Quantity 10/Pk.