Learn More
Thermo Scientific™ Remel™ BHI Agar with Vancomycin
GSA_VA
Catalog No. r01176 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
Catalog No. Format
R01176 12mm x 85mm Monoplate
1 options
This item is not returnable. View return policy
Catalog No. R01176 Supplier Thermo Scientific™ Supplier No. R01176
Add to Cart
Add to Cart
This item is not returnable. View return policy
Detect vancomycin-resistant enterococci with this ready-to-use medium recommended by CLSI.
Detect vancomycin-resistant enterococci with Thermo Scientific™ Remel™ Brain Heart Infusion (BHI) Agar with Vancomycin. Brain Heart Infusion (BHI) Agar with Vancomycin is used for screening of enterococci resistant to vancomycin and is recommended by the Clinical and Laboratory Standards Institute (CLSI) as a reliable method for the detection of all phenotypes of vancomycin-resistant enterococci1,2.
Vancomycin-resistant enterococci were first recognized in Europe and the United States in the late 1980s. Since then they are now being recovered with increasing frequency3. Various phenotypes responsible for acquired resistance in enterococci have been described with high to low vancomycin resistance and cross-resistance to teicoplanin4. Detection has been difficult due to failure of commonly used susceptibility methods to identify those isolates with low-level resistance5.
An Agar screen test with BHI Agar supplemented with 6μg/ml of vancomycin has been recommended by the Clinical and Laboratory Standards Institute (CLSI) as a reliable method for the detection of all phenotypes of vancomycin-resistant enterococci1,2.
  • Recommended by the CLSI as a reliable method for the detection of all phenotypes of vancomycin-resistant enterococci1,2
  • Ready-to-use convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description BHI Agar w/Vancomycin 6mcg/mL; Monoplate
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.