Learn More
Thermo Scientific™ BHI Agar
GSA_VA
Catalog No. R01132 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
15mm x 103mm Tube
Monoplate Deep Pour
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
R01132 Monoplate Deep Pour
R01130 12mm x 85mm Monoplate
R060230 15mm x 103mm Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R01132 Supplier Thermo Scientific™ Supplier No. R01132
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Cultivate a wide variety of microorganisms including bacteria, yeasts, and molds with the nutritious, ready-to-use base medium.
Cultivate a wide variety of microorganisms including bacteria, yeasts, and molds with the nutritious base medium Thermo Scientific™ Remel™ Blood Heart Infusion (BHI) Agar with Sheep Blood1.
Use BHI Agar for the cultivation of a wide variety of microorganisms, including pathogenic fungi.
  • Highly Nutritious: medium recommended for the enrichment of fastidious organisms.
  • Ready-to-use: the convenience of prepared media.
BHI Agar contains brain heart infusion, peptone, and dextrose, which provide nitrogen, carbon, sulfur, vitamins and a carbohydrate source. Sheep blood provides essential growth factors for fastidious fungi. Yeast extracts are incorporated in the medium to serve as a growth stimulant. BHI Agar can also be used for: Cryptococcus neoformans ATCC® 14116, Escherichia coli ATCC® 25922 and Staphylococcus aureus ATCC® 25923.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: BAM, COMPF, EPA, IDF, SMD.
Description BHI Agar (Brain Heart Infusion Agar)
Format Monoplate Deep Pour
Product Type Agar Plate
Quantity 10/Pk.