Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ BHI Broth (Brain Heart Infusion Broth)
GSA_VA

Catalog No. R060258 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 x 0.5 mL
100 x 2 mL
100 x 5 mL
10 x 500 mL
Format:
15mm x 103mm Tube
Borosilicate Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R060258 100 x 2 mL 15mm x 103mm Tube
R060270 100 x 5 mL 15mm x 103mm Tube
R112022 10 x 500 mL Borosilicate Bottle
R14101 100 x 0.5 mL 15mm x 103mm Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. R060258 Supplier Thermo Scientific™ Supplier No. R060258
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ Remel BHI Broth (Brain Heart Infusion Broth) is for the cultivation of microorganisms.

Thermo Scientific™ Remel BHI Broth (Brain Heart Infusion Broth) is for the cultivation of microorganisms.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM, SMD, SMWW, USDA.
Description BHI Broth
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 100 x 2 mL

REF R14101 for use with MicroScan™ System. REF R112022 not intended for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.