Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ BHI Broth w/Hemin, Vitamin K

Catalog No. R060278 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 5 mL
100 x 5 mL
Format:
15mm x 103mm Tube
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R060278 20 x 5 mL 15mm x 103mm Tube
R060276 100 x 5 mL 15mm x 103mm Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R060278 Supplier Thermo Scientific™ Supplier No. R060278
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ Remel BHI Broth with Hemin, Vitamin K (Brain Heart Infusion Broth) is for the cultivation of aerobic and anaerobic microorganisms.

Thermo Scientific™ Remel BHI Broth with Hemin, Vitamin K (Brain Heart Infusion Broth) is for the cultivation of aerobic and anaerobic microorganisms.

TRUSTED_SUSTAINABILITY

Specifications

Description BHI Broth w/Hemin; Vitamin K
Format 15mm x 103mm Tube
Product Type Microbiology Media
Packaging Type 20/Pack
Quantity 20 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.