Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Bile Esculin Azide Broth

Catalog No. R060302 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R060302 20 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R060302 Supplier Thermo Scientific™ Supplier No. R060302
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective medium is for the isolation and presumptive identification of group D streptococci and enterococci.

Thermo Scientific™ Remel Bile Esculin Azide Broth Selective medium is for the isolation and presumptive identification of group D streptococci and enterococci.

TRUSTED_SUSTAINABILITY

Specifications

Description Bile Esculin Azide Broth
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 20 x 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.