Description
Thermo Scientific™ Remel bio-spo Sponge is a sterile cellulose sponge pre-moistened with 10mL Neutralizing Buffer.
- Individually wrapped in foil pouch
- Manufacturer: Solar Biologicals
Ordering Alerts:
Individually wrapped in foil pouch.
Specifications
Specifications
| Description | bio-spo Sponge w/Neutralizing Buffer |
| Quantity | 100/Pk. |
| Volume (Metric) | 10 mL |