Learn More
Thermo Scientific™ Remel™ Biplates, HE through V Agar, HE Agar, XLD Agar
Catalog No. R02086 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
R02086 10/Pk.
1 options
This item is not returnable. View return policy
Catalog No. R02086 Supplier Thermo Scientific™ Supplier No. R02086
Add to Cart
Add to Cart
This item is not returnable. View return policy
Perform screening and isolation in the same 12 x 85mm dish, divided into two sections to hold either the same media on both sides or two media, one on each side.
Perform screening and isolation in the same 12 x 85mm dish using Thermo Scientific™ Remel Biplates, divided into two sections to hold either the same media on both sides or two media, one on each side.
Salmonella and Shigella are major causes of bacterial enteric illness with transmission occurring via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water.
  • Ready-to-use <
TRUSTED_SUSTAINABILITY
Specifications
Description HE Agar; XLD Agar
Format Biplate
Product Type Agar Plate
Quantity 10/Pk.