Learn More
Description
Selectively isolate Streptococcus pneumoniae from clinical specimens with this selective ready prepared medium.
Selectively isolate Streptococcus pneumoniae from clinical specimens with Thermo Scientific™ Remel™ Blood Agar with Gentamicin. The addition of 5% sheep blood enriches the medium by providing the essential growth factors and allows hemolytic rections to be observed. Gentamicin is a selective agent inhibitory to most Enterobacteriaceae, Neisseri, and Staphylococcus spp.
Streptococcus pneumoniae is a common cause of otitis media, an infection of the ear canal. Due to increased resistance to antimicrobial agents, recovery of S.pneumoniae in culture and subsequent antimicrobial susceptibility testing has become essential. Blood Agar with Gentamicin allows recovery of isloated colonies and allows initiation of confirmatory procedures from the primary isolation plate.
- Selective: Gentamicin is inhibitory to most Enterobacteriaceae, Neisseri, and Staphylococcus
- Ready-to-use: convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Description | Blood Agar w/Gentamicin (TSA w/5% Sheep Blood) |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.