Learn More
Description
Selectively isolate and differentiate Salmonella species other than Salmonella typhi from food and other materials with this selective medium.
Selectively isolate and differentiate Salmonella species other than Salmonella typhi from food and other materials with Thermo Scientific™ Remel™ Brilliant Green Agar with Novobiocin. The medium is recommended by the American Public Health (APHA) for the recovery of Salmonella from food products1. Moats added novobiocin to the medium to inhibit growth of Proteus in culture from beef and poultry products2.
Salmonella infection (salmonellosis) is a common bacterial disease that affects the intestinal tract. Salmonella bacteria typically live in animal and human intestines and are shed through feces. Humans become infected most frequently through contaminated water or food.
Use Brilliant Green Agar for isolation of salmonellae other than Salmonella typhi.
- Selective: brilliant green in the medium inhibits the growth of non-target organisms.
- Easy-to-read: Salmonella colonies are easy to read as they range from reddish to pink to nearly white in color with red zone.
- Corresponds to the medium recommended by the APHA1
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Description | Brilliant Green Agar w/Novobiocin |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |
REF R110113 not intended for IVD use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.