Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Brilliant Green Bile Broth 2%

Catalog No. R07022 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R07022 20 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R07022 Supplier Thermo Scientific™ Supplier No. R07022
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ Remel Brilliant Green Bile Broth 2% is for the detection of coliform microorganisms in water, wastewater, foods, and dairy products.

Thermo Scientific™ Remel Brilliant Green Bile Broth 2% is for the detection of coliform microorganisms in water, wastewater, foods, and dairy products.

Fermentation is detected by gas in Durham tube.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, EPA, IDF, SMWW.
Description Brilliant Green Bile Broth 2%
Format 16mm x 125mm Durham Tube
Product Type Microbiology Media
Quantity 20 x 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.