Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Brucella Agar with 5% Sheep Blood, Hemin and Vitamin K
GSA_VA

Catalog No. R01255 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
Format:
12mm x 85mm Monoplate
15mm x 150mm Plate
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R01255 100/Pk. 12mm x 85mm Monoplate
R04011 10/Pk. 15mm x 150mm Plate
R01254 10/Pk. 12mm x 85mm Monoplate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R01255 Supplier Thermo Scientific™ Supplier No. R01255
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate and cultivate anaerobic bacteria including Brucella, from clinical and non-clinical specimens with this ready-to-use formulation that conforms to APHA.

Isolate and cultivate anaerobic bacteria including Brucella, from clinical and non-clinical specimens with Thermo Scientific™ Remel™ Brucella Agar prepared plates with 5% Sheep Blood, Hemin and Vitamin K. Brucella Agar with 5% Sheep Blood, Hemin and Vitamin K is prepared according to the formulation of the American Health Association (APHA)1. Finegold reported Brucella agar with 5% sheep blood is preferable to heart infusion agar for the cultivation of anaerobic bacteria2. The addition of Vitamin K, hemin and sheep blood in the medium makes it appropriate for the cultivation of many anaerobes.

Brucellosis is a highly contagious zoonosis caused by ingestion of unpasteurized milk or undercooked meat from infected animals, or close contact with their secretions.

Brucella Agar is developed for the cultivation of Brucella species from diagnostic specimens such as blood, and from foods and potentially contaminated material.

  • Prepared according to the formulation of the American Health Association (APHA)1.
  • Preferable to Heart Infusion Blood Agar Base for the cultivation of anaerobic bacteria2.
  • Ready-to-use convenience of prepared media.

Brucella Agar with 5% Sheep Blood, Hemin and Vitamin K is a rich nutrient base that supports the growth of fastidious organisms.  The media contains dextrose which provides an energy source; peptones to provide nitrogenous compounds, and yeast extract to supply B vitamins. Growth factors required by some anaerobic bacteria are provided for in the added sheep blood, which also allows the demonstration of hemolytic reactions. Hemin and vitamin K are incorporated into the medium to enhance the growth of gram-positive spore-formers and Bacteroides species.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Brucella Agar w/5% Sheep Blood, Hemin, Vitamin K
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.