Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Brucella Agar with 5% Sheep Blood, TVP (Skirrow)

Catalog No. R01258 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
R01258 12mm x 85mm Monoplate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R01258 Supplier Thermo Scientific™ Supplier No. R01258
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate anaerobic microorganisms with this ready prepared medium prepared according to American Public Health Association.

Isoate and cultivate Campylobacter jejuni from fecal specimens and rectal swabs with Thermo Scientific™ Remel™ Brucella Agar with 5% Sheep Blood, TVP (Skirrow). Campylobacter is a major cause of diarrheal disease in adults and children and usually is associated with unsanitary preparation of food.

The medium based on Skirrow's formulation contains three antibiotics: trimethorprim, vancomycin and polymyxin B. In order to effectively recover Campylobacter spp without comprising the activity of trimethorprim, horse blood in the original formulation is substituted by the sheep blood.

Campylobacter are Gram-negative, spirally shaped microaerophilic organisms which may be present in raw milk, untreated water, improperly handled food and undercooked meats, poultry and shellfish. Human consumption of these organisms can result in a range of clinical illnesses from transient asymptomatic colonisation to severe dysentery. The symptoms of Campylobacter enteritis include diarrhoea, stomach pain, nausea, fever, headache and muscle pain.

Brucella Agar with 5% Sheep Blood, TVP (Skirrow) is a solid medium recommended for use in qualitative procedures for the primary isolation and cultivation of Campylobacter jejuni from fecal specimens or rectal swabs.

  • Selective:  due to the presence of the selective agents like trimethorprim, vancomycin and polymyxin B the medium is selective for Campylobacter
  • Ready-to-use: convenicne of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Brucella Agar w/5% Sheep Blood, Trimethoprim, Vancomycin, Polymyxin B
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.