Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Brucella Agar with 5% Sheep Blood
GSA_VA

Catalog No. R01252 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
R01252 12mm x 85mm Monoplate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R01252 Supplier Thermo Scientific™ Supplier No. R01252
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Isolate and cultivate fastidious aerobic as well as anaerobic microorganisms with this medium.

Isolate and cultivate fastidious aerobic as well as anaerobic microorganisms with Thermo Scientific™ Remel™ Brucella Agar with 5% Sheep Blood. It is prepared according to the formulation of the American Public Health Association for Albimi Broth1,2 and recommended for isolation obligate and facultative anaerobes from clinical specimens3,4.

Casein and meat peptones in the medium supply nitrogen, amino acids, and peptides necessary for bacterial growth.Dextrose is a ready source of energy, Yeast extract provides B-complex vitamins and enhances growth. Sodium chloride us a sorce of essential electrolytes and maintains osmotic equilibrium. Sodium bisulfite is a reducing agent. Sheep blood provides growth factors required by certain anaerobic organisms and allows for demonstration of hemolysis by aerobic and anerobic bacteria. Agar is a solidifying agent.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Brucella Agar w/5% Sheep Blood
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.