Learn More
Thermo Scientific™ Brucella Agar with Laked blood, Hemin and Vitamin K
Shop All Thermo Scientific Products
Description
Isolate anaerobic micro-organisms with Thermo Scientific™ Remel™ Brucella Agar with Laked blood, Hemin and Vitamin K. Prepared according to American Public Health Association1 the medium contains laked blood and supports the growth of fastidious anaerobic bacteria. The medium also contains menadione (Vitamin K) and hemin for added enrichment to enhance the growth of anerobic bacteria. Brucella Agar with Laked blood, Hemin and Vitamin K can also be used for antimicrobial susceptibility testing of anaerobes2,3
Use Brucella Agar with Laked blood, Hemin and Vitamin K in qualitative procedures for the isolation of anaerobic bacteria.
- Prepared according to American Public Health Association1
- Can can also be used for antimicrobial susceptibility testing of anaerobes2,3
- Ready to use convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Brucella Agar w/Laked Blood, Hemin, Vitamin K |
| Format | 15mm x 150mm Plate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |