Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Brucella Broth

Catalog No. p-4523801
Change view
Click to view available options
Quantity:
10 x 5 mL
20 x 5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R060320 10 x 5 mL
R060322 20 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R060320 Supplier Thermo Scientific™ Supplier No. R060320
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Thermo Scientific™ Remel Brucella Broth is for the cultivation of Brucella and other fastidious microorganisms.

Thermo Scientific™ Remel Brucella Broth is for the cultivation of Brucella and other fastidious microorganisms.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: USDA.
Description Brucella Broth
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 10 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.