Learn More
Thermo Scientific™ Remel™ Buffered CYE Differential Agar w/BTB, BCP, Polymyxin B, Anisomycin, Vancomycin
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Remel Buffered CYE Differential Agar BTB, BCP, Polymyxin B, Anisomycin, Vancomycin (BCYE wDyes PAV) is for isolation and differentiation of Legionella spp.
- Contains dyes to differentiate Tatlockia micdadei from Legionella pneumophila and antibiotics to suppress contaminating flora
Specifications
Specifications
| Description | Buffered CYE Differential Agar |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.