Learn More
Thermo Scientific™ Remel™ Buffered CYE Differential Agar w/BTB, BCP
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Remel Buffered CYE Differential Agar w/BTB, BCP (BCYE w/Dyes) is used for qualitative procedures to isolate Legionella species.
- Contains dyes to differentiate Tatlockia spp.from Legionella pneumophila
Specifications
Specifications
| Description | Buffered CYE Differential Agar w/BTB, BCP |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.