Learn More
Thermo Scientific™ Remel™ Buffered CYE Selective Agar w/Dyes, Glycine, Vancomycin, Polymycin B
Shop All Thermo Scientific Products
Description
Thermo Scientific™ Remel Buffered CYE Select Agar w/Dyes, Glycine, Vancomycin,Polymycin B(BCYE w/DGVP) is used for selective and differential isolation of Legionella spp. from clinical specimens and environmental samples.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Buffered CYE Selective Agar w/Dyes, Glycine, Vancomycin, Polymycin B |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |