Learn More
Thermo Scientific™ Remel™ Buffered CYE Selective Agar w/Glycine, Polymyxin B, Anisomycin, Vancomycin
Catalog No. R110107 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
Monoplate
Catalog No. Format
R110107 Monoplate
1 options
This item is not returnable. View return policy
Catalog No. R110107 Supplier Thermo Scientific™ Supplier No. R110107
Add to Cart
Add to Cart
This item is not returnable. View return policy
Use for selective recovery of Legionella pneumophila while reducing contaminating microorganisms from potable water samples.
Thermo Scientific™ Remel Buffered CYE Select Agar Glycine, Polymyxin B, Anisomycin, Vancomycin BCYE Glycine PAV is for selective recovery of Legionella pneumophila while reducing contaminating microorganisms from potable water samples.
TRUSTED_SUSTAINABILITY
Specifications
Description Buffered CYE Selective Agar w/Glycine, Polymyxin B, Anisomycin, Vancomycin
Format Monoplate
Product Type Agar Plate
Quantity 10/Pk.
Not intended for IVD use.