Learn More
Thermo Scientific™ Remel™ Buffered CYE Selective Agar w/Polymyxin B, Anisomycin, Cefamandole
Shop All Thermo Scientific Products
Description
Thermo Scientific™ Remel Buffered CYE Selective Agar w/Polymyxin B, Anisomycin, Cefamandole BCYE w/PAC is used for selective isolation of Legionella spp.
- Cefamandole is inhibitory for Tatlockia micdadei
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Buffered CYE Selective Agar; w/Cefamandole |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |
Nonselective BCYE, REF R01342, is recommended for use with this medium.