Learn More
Thermo Scientific™ Remel™ Buffered CYE Selective Agar w/Polymyxin B, Anisomycin, Vancomycin
GSA_VA
Catalog No. R01341 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
Catalog No. Format
R01341 12mm x 85mm Monoplate
1 options
This item is not returnable. View return policy
Catalog No. R01341 Supplier Thermo Scientific™ Supplier No. R01341
Add to Cart
Add to Cart
This item is not returnable. View return policy
Use for selective isolation of Legionella spp.
Thermo Scientific™ Remel Buffered CYE Selective Agar wPolymyxin B, Anisomycin, Vancomycin (BCYE w/PAV) is used for selective isolation of Legionella spp.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Buffered CYE Selective Agar; w/Vancomycin
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.
Nonselective BCYE, REF R01342, is recommended for use with this medium.