Learn More
Thermo Scientific™ Campy Blood Agar Blaser with 5% Sheep Blood and Antibiotics
Shop All Thermo Scientific Products

Description
Carry out primary and selective isolation of Campylobacter species from fecal specimens with this selective, ready-to-use medium.
Carry out primary and selective isolation of Campylobacter species from fecal specimens with the Thermo Scientific™ Remel™ Campy Blood Agar Blaser with 5% Sheep Blood and Antibiotics. Campy Blood Agar Blaser with 5% Sheep Blood and Antibiotics is a modification of Brucella Agar Base with Sheep blood and Antibiotics by Blaser. The additional antibiotic cephalothin was added to improve the inhibition of commensal microbial flora in fecal specimens1.
Campylobacter are Gram-negative, spirally shaped microaerophilic organisms which may be present in raw milk, untreated water, improperly handled food and undercooked meats, poultry and shellfish. Human consumption of these organisms can result in a range of clinical illnesses from transient asymptomatic colonization to severe dysentery. The symptoms of Campylobacter enteritis include diarrhea, stomach pain, nausea, fever, and headache and muscle pain.
Use Campy Blood Agar with 5% Sheep Blood, Antibiotics for primary and selective isolation of Campylobacter from fecal specimens.
- Selective: the incorporation of cephalothin, amphotericin B, trimethoprim, vancomycin, and polymyxin B in the medium makes the medium inhibitory to Enterobacteriaceae, staphylococci, and yeast.
- Ready to use: convenience of prepared media.
Casein and meat peptones in the medium provide nitrogen, carbon, and sulfur which are essential for the recovery of Campylobacter sp. Sodium is a source of essential electrolytes and maintains osmotic equilibrium. Dextrose is an energy source and yeast extract supplies B vitamins. Sheep blood provides hemin and other necessary growth factors.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Campy Blood Agar Blaser |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.