Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Campy-Cefex Agar

Catalog No. R110138 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
R110138 12mm x 85mm Monoplate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R110138 Supplier Thermo Scientific™ Supplier No. R110138
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selectively isolate and differentiate Camplylobacter species from food and poultry industry with this selective medium

Selectively isolate and differentiate Camplylobacter species from food and poultry industry with Thermo Scientific™ Remel™ Campy-Cefex Agar. The Agar is recommended by the Natonal Advisory Committee on Microbiological Criterion on Food (NACMCF) for isolation of Campylobacter spp. from poultry1.Campy Cefex Agar is also recommended by American Public Health Association (APHA) and United States Department of Agriculture (USDA) for the isolation of Campylobacter spp. from foods and poultry2,3

Campylobacter is a major cause of acute diarrheal disease in humans4. Consumption of contaminated food and water accounts for 70% of Campylobacter related illnesses each year with poultry identified as a primary reservoir.

Campy Cefex Agar is a solid medium recommended for use in qualitative procedures for selective and differential isolation of Camylobacter species from food and poultry.

  • Selective: cefoperazone is a selective agent which inhibits commensal microbial fecal flora and cycloheximide inhibits the growth of fungi.
  • Ready-to- use: convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Campy Cefex Agar
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.

Not for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.