Learn More
Thermo Scientific™ Campylobacter Selective Medium (leaked horse blood, vancomycin, polymyxin B, trimethoprim, cefaperazone)
GSA_VA
Catalog No. R01274 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
Catalog No. Format
R01274 12mm x 85mm Monoplate
1 options
This item is not returnable. View return policy
Catalog No. R01274 Supplier Thermo Scientific™ Supplier No. R01274
Add to Cart
Add to Cart
This item is not returnable. View return policy
Selectively isolate Campylobacter species from fecal specimens with this selective ready to use medium.
Selectively isolate Campylobacter species from fecal specimens with Thermo Scientific™ Remel™ Campylobacter Selective Medium (leaked horse blood, vancomycin, polymyxin B, trimethoprim, cefaperazone). The medium is a modification of original Skirrow formula with the addition of 50 mg/l of cefoperazone. Cefoperazon is a cephalosporin antibiotic with enhanced activity against pseudomonas and members of the Enterobacteriaceae.
Campylobacter jejuni is a major cause of acute diarrheal diseae in humans1. It has worldwide distribution, and is ubiquitous in domestic pets and food animals.
Campylobacter Selective Agar is supplemented with a range of antimicrobials to inhibit background flora (including vancomycin, polymyxin B, trimethoprim, cefaperazone).
  • Laked horse blood supplies X factor (hemin) and other necessary growth factors
  • Vancomycin inhibits Gram-positive bacteria, polymyxin B inhibits most Gram negative bacilli, and trimethoprim inhibits Proteus
  • Cefoperazone enhances suppression of Pseudomonas, thereby improving recovery of Campylobacter spp. from fecal specimens
  • Ready-to-use convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Campylobacter Selective Medium
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.