Description
Thermo Scientific™ Remel Casein Agar (Casein Hydrolysis Agar) is for the differentiation of aerobic actinomycetes.
Ordering Alerts:
10 tubes of Casein A and 10 tubes of Casein B per pkg. Mix together for pour plate.
Specifications
Specifications
| Description | Casein Agar (Casein Hydrolysis Agar) |
| Format | 20mm x 113mm Pour Tube |
| Product Type | Agar |
| Quantity | 20/Pk. |