Learn More
Thermo Scientific™ Remel™ CCFA Modified Agar w/Horse Blood (with cefoxitin, cycloserine, fructose)
Description
Thermo Scientific™ Remel CCFA Modified Agar w/Horse Blood (with cefoxitin, cycloserine, fructose) is used for selective isolation and differentiation of Clostridium difficile.
Specifications
Specifications
| Description | CCFA Modified Agar w/Horse Blood w/cefoxitin, cycloserine, fructose |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar |
| Quantity | 10/Pk. |