Learn More
Thermo Scientific™ Remel™ Charcoal Selective Medium (w/vancomycin, cycloheximide, cefoperazone)
GSA_VA
Catalog No. R01294 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
Catalog No. Format
R01294 12mm x 85mm Monoplate
1 options
This item is not returnable. View return policy
Catalog No. R01294 Supplier Thermo Scientific™ Supplier No. R01294
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Use for the selective isolation of Campylobacter species.
Thermo Scientific™ Remel Charcoal Selective Medium (w/vancomycin, cycloheximide, cefoperazone) is used for the selective isolation of Campylobacter species.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Charcoal Selective Medium
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.