Learn More
Thermo Scientific™ Remel™ Chocolate Agar w/Yeast Extract, Hemin, Vitamin K
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Remel Chocolate Agar w/Yeast Extract, Hemin, Vitamin K is used for the cultivation of fastidious microorganisms, particularly Haemophilus spp. and pathogenic Neisseria spp.
Specifications
Specifications
| Description | Chocolate Agar with Yeast Extract |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |