Learn More
Thermo Scientific™ Remel™ Clostridium Difficile Agar (w/cefoxitin, cycloserine, fructose, egg yolk)
Catalog No. R01310 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
Catalog No. Format
R01310 12mm x 85mm Monoplate
1 options
This item is not returnable. View return policy
Catalog No. R01310 Supplier Thermo Scientific™ Supplier No. R01310
Add to Cart
Add to Cart
This item is not returnable. View return policy
Use for selective isolation and differentiation of Clostridium difficile.
Thermo Scientific™ Remel Clostridium Difficile Agar (w/cefoxitin, cycloserine, fructose, egg yolk) is used for selective isolation and differentiation of Clostridium difficile.
TRUSTED_SUSTAINABILITY
Specifications
Description Clostridium Difficile Agar
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.