Learn More
Thermo Scientific™ Remel™ Color Slide™ II Mononucleosis Test
Description
Thermo Scientific™ Remel Color Slide and II Mononucleosis Test is a rapid slide agglutination test using horse erythrocytes for detection of heterophile antibodies of infectious mononucleosis.
- Dual color indicator
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Color Slide™ II Mononucleosis Test |
| Detectable Analytes | Mononucleosis Antibodies |
| Sample Type | Plasma, Serum |
| Type | Reagent |
| Quantity | 150 Tests |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.