Learn More
Thermo Scientific™ Remel™ Columbia Horse Blood Bi-Layer Medium
Catalog No. R01477 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
Catalog No. Format
R01477 12mm x 85mm Monoplate
1 options
This item is not returnable. View return policy
Catalog No. R01477 Supplier Thermo Scientific™ Supplier No. R01477
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel Columbia Horse Blood Bi-Layer Medium is used for isolation and identification of Listeria monocytogenes.
Thermo Scientific™ Remel Columbia Horse Blood Bi-Layer Medium is used for isolation and identification of Listeria monocytogenes.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: USDA.
Description Columbia Horse Blood Bi-Layer Medium
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.
Not intended for IVD use.