Learn More
Thermo Scientific™ Remel™ Contact I Potato Dextrose Agar
Catalog No. R111045 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
Contact - Single Bag
Catalog No. Format
R111045 Contact - Single Bag
1 options
This item is not returnable. View return policy
Catalog No. R111045 Supplier Thermo Scientific™ Supplier No. R111045
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel Contact I Potato Dextrose Agar is used for cultivation of yeast and fungi.
Thermo Scientific™ Remel Contact I Potato Dextrose Agar is used for cultivation of yeast and fungi.
  • Manufactured in a clean room environment to provide the highest level of sterility assurance
  • Made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
  • Contact I (Single Bag) and Contact II (Double Bag) are manufactured in a clean room environment
  • This provides the highest level of sterility assurance with no compromise of growth characteristics
  • Each lot is tested to ensure consistent, reproducible results
TRUSTED_SUSTAINABILITY
Specifications
Description Contact I Potato Dextrose Agar
Format Contact - Single Bag
Product Type Agar Plate
Quantity 20/Pk.
Not intended for IVD use.