Learn More
Thermo Scientific™ Remel™ Contact I Sabouraud Dextrose Agar (w/lecithin, polysorbate 80) (pH 5.6)
Catalog No. R111105 Shop All Thermo Scientific Products
This item is not returnable. View return policy
Catalog No. R111105 Supplier Thermo Scientific™ Supplier No. R111105
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel Contact I Sabouraud Dextrose Agar (w/lecithin, polysorbate 80) (pH 5.6) is used for cultivation of yeast and fungi.
Thermo Scientific™ Remel Contact I Sabouraud Dextrose Agar (w/lecithin, polysorbate 80) (pH 5.6) is used for cultivation of yeast and fungi.
  • Manufactured in a clean room environment to provide the highest level of sterility assurance
  • Made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
  • Contact I (Single Bag) and Contact II (Double Bag) are manufactured in a clean room environment
  • This provides the highest level of sterility assurance with no compromise of growth characteristics
  • Each lot is tested to ensure consistent, reproducible results
TRUSTED_SUSTAINABILITY
Specifications
Description Contact I Sabouraud Dextrose Agar
Format Contact - Single Bag
Product Type Agar Plate
Quantity 20/Pk.
Not intended for IVD use.