Learn More
Thermo Scientific™ Remel™ Contact I Tryptic Soy Agar w/Lecithin, Polysorbate 80
GSA_VA
Catalog No. R111101 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R111101 20/Pk.
R111102 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R111101 Supplier Thermo Scientific™ Supplier No. R111101
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel Contact I Tryptic Soy Agar w/Lecithin, Polysorbate 80 is used for the cultivation of bacteria, yeast and fungi.
  • Manufactured in a clean room environment to provide the highest level of sterility assurance with no compromise of growth characteristics
  • Each lot is tested to ensure consistent, reproducible results
  • Made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Contact I Tryptic Soy Agar w/Lecithin, Polysorbate 80
Format Contact - Single Bag
Product Type Agar Plate
Quantity 20/Pk.
Not intended for IVD use.