Description
Thermo Scientific™ Remel Contact I Tryptic Soy Agar is used for the cultivation of bacteria, yeast and fungi.
- Contact I (Single Bag) and Contact II (Double Bag) are manufactured in a clean room environment
- This provides the highest level of sterility assurance with no compromise of growth characteristics
- Each lot is tested to ensure consistent, reproducible results
- Made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
Specifications
Specifications
| Description | Contact I Tryptic Soy Agar |
| Format | Contact - Single Bag |
| Product Type | Agar Plate |
| Quantity | 20/Pk. |
Not intended for IVD use.